Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Trypticase™ Soy Agar w/Lecithin and Polysorbate 80 Sterile Pack Media - RODAC™ Plates

Catalog No. B4392335 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
TSA, w/Lecithin and Polysorbate 80, RodacPlate; 100/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
B4392335 TSA, w/Lecithin and Polysorbate 80, RodacPlate; 100/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B4392335 Supplier BD Supplier No. 292335

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gamma-irradiated plated media individually wrapped, then double wrapped in Tyvek™/ polyethylene bags

Rodac (Replicate Organism Detection and Counting) plates for surface sampling.

  • For critical environments

Compliance

Tested for performance and sterility according to Association for the Advancement of Medical Instrumentation (AAMI) guidelines.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AAMI
Color Light Beige
Content And Storage 2°C to 8°C
Format Rodac
Manufacturing Origin United States
Media Type Trypticase Soy Agar w/Lecithin and Polysorbate 80
Sterility Sterile
Formulation Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Lecithin 0.7g/L, Polysorbate 80 5g/L, Agar 15g/L
For Use With (Application) Used for microbial load testing
Includes Inner bag for transportation
Packaging Type Plate
Quantity 100/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.