Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ VDRL Test Control Serum Set

Catalog No. DF3520327 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF3520-32-7 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Contains dry natural rubber and sodium azide.
Catalog No. DF3520-32-7 Supplier BD Supplier No. 235201

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Contains dry natural rubber and sodium azide.

Includes

Nontreponemal Antigen Reactive Serum (1 vial); VDRL Weakly Reactive Serum (1 vial); Nontreponemal Antigen Nonreactive Serum (1 vial); Aliquant Vials (3 vials)

Used in quality control testing of VDRL Antigen by the slide flocculation test

Treponema pallidum is the causative agent of syphilis. Syphilis is a chronic infection with many clinical manifestations which occur in distinct stages. Specific laboratory tests are recommended for the detection of each stage of the disease.

  • No cross reactivity
  • Highly specific
  • Fast reaction times
  • Easy, clear results
  • Rehydrate to 3mL per vial

TRUSTED_SUSTAINABILITY

Specifications

Detectable Analytes T. pallidum
Type VDRL Test Control Serum Set
Formulation Cardiolipin 0.03% and Cholesterol 0.9% dissolved in absolute Alcohol with sufficient Lecithin (∼0.20%) to produce standard reactivity
For Use With (Application) Used in quality control testing of VDRL Antigen by the slide flocculation test
Includes Nontreponemal Antigen reactive Serum 1 vial, VDRL weakly reactive Serum 1 vial, nontreponemal Antigen nonreactive Serum1 vial, aliquant vials 3 vials
Quantity Each

For in vitro Diagnostic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.