Learn More
Cole-Parmer B-D Disposable Syringe, Non-Sterile, Luer-Lok, 50 mL, 25/Pk
Catalog No. NC1928263 Shop All Cole-Parmer Products
Encompass_Preferred
Use these latex-free, disposable polypropylene syringes for your general-purpose applications. Available in either Luer-Lok or slip-tip versions. Syringes come nonsterilized in bulk packages and can be autoclaved. Needles and shield tips are not included; order separately.
Catalog No. NC1928263 Supplier Cole-Parmer Supplier No. 0794428
Use these latex-free, disposable polypropylene syringes for your general-purpose applications. Available in either Luer-Lok or slip-tip versions. Syringes come nonsterilized in bulk packages and can be autoclaved. Needles and shield tips are not included; order separately.