Learn More
Cole-Parmer BD Biocoat Disposable Syringe, Non-Sterile, Slip-Tip, 5 mL, 100/Pk
Catalog No. NC1795794 Shop All Cole-Parmer Products
Encompass_Preferred
Use these latex-free, disposable polypropylene syringes for your general-purpose applications. Available in either Luer-Lok or slip-tip versions. Syringes come nonsterilized in bulk packages and can be autoclaved. Needles and shield tips are not included; order separately.
Catalog No. NC1795794 Supplier Cole-Parmer Supplier No. 0794408
Use these latex-free, disposable polypropylene syringes for your general-purpose applications. Available in either Luer-Lok or slip-tip versions. Syringes come nonsterilized in bulk packages and can be autoclaved. Needles and shield tips are not included; order separately.