Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Fixable Viability Stain 700

Catalog No. bdb564997 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564997 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564997 Supplier BD Supplier No. 564997
Only null left
Add to Cart
Add to Cart

For discrimination of viable from non-viable mammalian cells in multicolor flow cytometric applications. Reacts with and covalently binds to cell-surface and intracellular amines.

  • Permeable plasma cell membranes, such as those present in necrotic cells, allow intracellular diffusion of dye and covalent binding to higher overall concentrations of amines than in non-permeable live cells
  • Necrotic cells present in a typical in vitro assay label with higher levels of dye increases their fluorescence intensity 10-20 fold over viable cells
  • Labeled cells can be fixed with formaldehyde for downstream decontamination, freezing and/or permeabilization and subsequent intracellular staining while maintaining stable viability stain fluorescence
  • Excited by Red laser (with an excitation maximum of 657nm)
  • Fluorescence emission maximum of 700nm
  • Applications: Flow cytometry, Intracellular staining
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 μg
For Use With (Application) Flow cytometry, Intracellular staining (flow cytometry) (Tested During Development)

RUO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.