Learn More
BD Biotin Human CD4 T Lymphocyte Enrichment Set - DM
Catalog No. BDB557939 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 109 Cells
Catalog No. Quantity
BDB557939 1 x 109 Cells
1 options
Catalog No. BDB557939 Supplier BD Supplier No. 557939
Add to Cart
Add to Cart
For the negative selection of CD4 T lymphocytes from peripheral blood
Magnetic Labeling
TRUSTED_SUSTAINABILITY
Specifications
Conjugate Biotin
Quantity 1 x 109 Cells
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Research Use Only