Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Biotin Human NK Cell Enrichment Set - DM

Catalog No. BDB557987 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 109 Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557987 1 x 109 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557987 Supplier BD Supplier No. 557987
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

CD3, CD19, CD36, CD41a, CD66b, CD123, CD235a, IgE biotinylated antibody cocktail and BD IMag SAv particles

Negative selection of Natural Killer (NK) cells from peripheral blood

Set consists of Biotinylated Human NK Cell Enrichment Cocktail and BD IMag™ Streptavidin Particles Plus

  • Reactivity: Human
  • Format: BD IMag-DM, Biotin

Magnetic Labeling

TRUSTED_SUSTAINABILITY

Specifications

Conjugate Biotin
Quantity 1 x 109 Cells

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.