Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Biotin Human T Lymphocyte Enrichment Set - DM

Catalog No. BDB557874 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557874 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557874 Supplier BD Supplier No. 557874
Add to Cart
Add to Cart

Includes

CD11b/Mac-1, CD16, CD19,CD36, CD41a, CD56, CD235a biotinylated antibody cocktail and BD IMag™ SAv particles

For negative selection of T lymphocytes from peripheral blood

The set components are Biotin Human T Lymphocyte Enrichment Cocktail and Biotin Human T Lymphocyte Enrichment Cocktail

Components

  • Biotin Human T Lymphocyte Enrichment Cocktail
  • Biotin Human T Lymphocyte Enrichment Cocktail

  • Reactivity: Human

Magnetic Labeling

TRUSTED_SUSTAINABILITY

Specifications

Cross Reactivity Human
Includes CD11b/Mac-1, CD16, CD19,CD36, CD41a, CD56, CD235a Biotinylated Antibody cocktail and BD IMag™ SAv particles
Product Type T-Lymphocyte Enrichment
Content And Storage 4°C
Form Magnetic Nanoparticles
For Use With (Application) Magnetic labeling
Quantity 5 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.