Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Biotin Mouse Dendritic Cell Enrichment Set - DM

Catalog No. BDB557955 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 x 109 Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557955 1 x 109 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557955 Supplier BD Supplier No. 557955
Add to Cart
Add to Cart

Includes

CD2, CD3e, CD45R/B220, CD49b, CD147, LY-6G and LY-6C, TER-119/Erythroid cells biotinylated antibody cocktail and BD IMag™ SAv particles

For the negative selection of dendritic cells (DC) from mouse spleen or lymph node

  • Reactivity: Mouse
  • Storage buffer: Aqueous buffered solution containing BSA and 0.09% sodium azide

Magnetic Labeling

TRUSTED_SUSTAINABILITY

Specifications

Conjugate Biotin
Quantity 1 x 109 Cells

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.