Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Biotin Mouse Hematopoietic Progenitor (Stem) Enrichment Set - DM

Catalog No. BDB558451 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
2 x 109 Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558451 2 x 109 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558451 Supplier BD Supplier No. 558451
Add to Cart
Add to Cart

Includes

Enrichment Cocktail (5mL): biotinylated monoclonal antibodies to mouse CD3ε (CD3 chain), CD11b (Integriα M chain), CD45R/B220, Ly-6G and Ly-6C (Gr-1) and TER-119/Erythroid Cells (Ly-76), SAv particles (5mL)

For immunomagnetic enrichment of hematopoietic progenitors

  • Optimized and pre-diluted to provide maximum efficiency
  • Reacts with cells from major hematopoietic cell lineages such as T lymphocytes, B lymphocytes, monocytes/macrophages, granulocytes and erythrocytes
  • Storage buffer comprises of aqueous buffered solution containing BSA and ≤0.09% sodium azide

Magnetic Labeling

TRUSTED_SUSTAINABILITY

Specifications

Conjugate Biotin
Quantity 2 x 109 Cells
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.