Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Bottled Media Saboraud Dextrose Agar

Catalog No. L99098 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Includes:
Chloramphenicol
No Chloramphenicol
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
L99098 Chloramphenicol
B4399510 No Chloramphenicol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. L99098 Supplier BD Supplier No. 299098
Add to Cart
Add to Cart
This item is not returnable. View return policy

For cultivation and maintenance of pathogenic and non-pathogenic fungi

  • Volume: 500mL
  • With and without Chloramphenicol

TRUSTED_SUSTAINABILITY

Specifications

Cap Type Screw Cap
Certifications/Compliance USP
Form Liquid
Format Bottle
Manufacturing Origin United States
Fill Volume 500 mL
Formulation Sabouraud Detrose Agar
For Use With (Application) Used for qualitative procedures for cultivation of pathogenic and nonpathogenic fungi, particularly dermatophytes
Includes Chloramphenicol
Packaging Type Bottle
Product Line BBL
Quantity 500 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.