Learn More
Description
- Ready-for-use prepared bottled media
- Helps detect microorganisms from pharmaceutical, food, environmental or water testing samples
- Used for the sterility testing of biologics and for the cultivation of anaerobes, aerobes, and microaerophiles
- Conforms with specifications of The United States Pharmacopeia (USP)
- Recommended in the FDA Bacteriological Analytical Manual (BAM) and the Official Methods of Analysis of AOAC International for food examination
- Specified for sterility checks on banked blood
- Contains sodium thioglycollate and L-cystine as reducing agents to prevent peroxide accumulation, inactivate heavy metal compounds, and maintain a low redox potential
- Resazurin acts as an oxidation-reduction indicator, appearing pink when oxidized and colorless when reduced
- Small amount of agar stabilizes the medium against convection currents
Specifications
Specifications
| Broth Type | Fluid Thioglycollate Medium |
| Certifications/Compliance | USP |
| Country of Origin | United States |
| Form | Liquid |
| Format | Bottle |
| Manufacturing Origin | United States |
| Media Type | Fluid Thioglycollate Medium |
| Preparation Method | Ready-to-Use |
| Product Type | Culture Medium |
| Fill Volume | 400 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.