Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Bottled Media TSB Lecithin

Catalog No. 14959801 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-959-801 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-959-801 Supplier BD Supplier No. 215187
Add to Cart
Add to Cart
This item is not returnable. View return policy

General non-selective growth media for a wide variety of organisms with disinfectant neutralization.

  • Intended for laboratory use as a general non-selective growth media for a wide variety of organisms, when the neutralization of disinfectants is necessary.
  • Shelf Life (Days): 365
  • For optimal growth of strict aerobes, containers should be vented during the incubation, which may be achieved by loosening the caps.
TRUSTED_SUSTAINABILITY

Specifications

Broth Type Tryptic Soy Broth
Country of Origin United States
Additive Polysorbate 80
Detects Microorganisms, Bacterial Growth, Fungal Growth
Form Liquid
Format Bottle
Manufacturing Origin United States
Media Type Broth
Product Type Culture Medium
Sterility Sterile
Test Type Growth Promotion Testing
Fill Volume 200 mL
Final Product Type Microbial Growth
Formulation Casein Peptone-17.0 g, Soy Peptone-3.0 g, Sodium Chloride-5.0 g, Dextrose-2.5 g, Polysorbate 80-5.0 g, Lecithin-0.7 g, Dipotassium Phosphate-2.5 g, Total-35.7 g/L
Incubation Conditions 20°C to 25°C
Packaging Type Bottle
Product Line BBL
Quantity 200 mL
Specimen Type Microbiological Samples
Protein Source Casein, Soy
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.