Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bovine serum albumin Stain Buffer, BD

Catalog No. BDB554657 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554657 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Highly toxic, dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing
Catalog No. BDB554657 Supplier BD Biosciences Supplier No. 554657
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Highly toxic, dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing

For immunofluorescent staining of single-cell suspensions

Useful for dilution and application of fluorescent reagents as well as for suspension, washing, and storage of cells destined for flow cytometric analysis

  • Buffered salt solution with neutral pH 7.4
  • Supplemented with 0.2% (w/v) bovine serum albumin proteins
  • Contains sodium azide which inhibits the potential redistribution of cell surface antigens caused by antibody crosslinking
  • Prevents potential loss of fluorescent signal intensities

Flow cytometry, Fluorescence microscopy, Intracellular staining (flow cytometry)

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Content And Storage 4°C
Format Liquid
Includes 0.2% BSA proteins and 0.09% sodium azide
Quantity 500mL
Validated Application RUO
Product Type Staining Buffer
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.