Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

embecta™ Alcohol Swabs

Catalog No. 1368063
Change view
Click to view available options
Composition:
70% Isopropyl Alcohol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Composition
13-680-63 70% Isopropyl Alcohol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
External Use Only
Catalog No. 13-680-63 Supplier embecta™ Supplier No. 326895

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
External Use Only

Isopropyl Alcohol 70% Alcohol Swab

  • Isopropyl Alcohol
  • 70% Alcohol Swab

Specifications

Product Type Alcohol Swab
Color White
Form Wipe
Composition 70% Isopropyl Alcohol
Disposable Single-use
For Use With (Application) Household Applications
Latex Free Yes
Solution Type 70% Isopropyl Alcohol
Sterility Non-sterile
Storage Requirements Controlled Room Temp

Made in the USA

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.