Learn More
Description
Includes
Fixation Buffer, Diluent Buffer, BDRetrievagen A antigen retrieval solution, biotinylated anti-BrdU antibody, ready-to-use Streptavidin-Horseradish peroxidase (Sav-HRP) solution, DAB Buffer, DAB Chromogen, and Control Slides
The BrdU In-Situ Detection Kit features our BD Retrievagen A, an antigen retrieval solution designed specifically to unmask antigenic sites, preserve tissue morphology, and enable simultaneous staining of other surface antigens in conjunction with BrdU.
- With BD Retrievagen A, an antigen retrieval solution
- Designed specifically to unmask antigenic sites, preserve tissue morphology, and enable simultaneous staining of other surface antigens in conjunction with BrdU
- Kit enables study of proliferation state of phenotypically defined cells within micro-environment of tissues
- Consistent results are assured by providing all critical reagents
- Comprehensive series of protocols for staining BrdU in paraffin-embedded and frozen tissue sections and cultured cells, and in conjunction with other antigens
- Control slides are provided in the kit to serve as a reference
Immunohistochemical staining
Specifications
Specifications
| Storage Requirements | 4°C |
| For Use With (Application) | Immunohistochemical Staining |
| Quantity | 1 Kit |
Safety and Handling
Research Use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.