Learn More
BD BrdU In-Situ Detection Kit
Catalog No. BDB550803 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
BDB550803 1 Kit
1 options
Highly toxic, avoid contact with skin, eyes, and mucous membranes
Catalog No. BDB550803 Supplier BD Supplier No. 550803
Add to Cart
Add to Cart
Highly toxic, avoid contact with skin, eyes, and mucous membranes
Includes
Fixation Buffer, Diluent Buffer, BD™ Retrievagen A Antigen Retrieval Solution, biotinylated anti-BrdU antibody, ready-to-use Streptavidin-Horseradish peroxidase (SAv-HRP) Solution, DAB Buffer, DAB Chromogen, and Control Slides
Requires
BrdU for in vitro or in vivo labeling, H2O2 (30%) solution for blocking endogenous peroxidase, phosphate buffer saline (PBS)
Determines proliferation rates and cell cycle kinetics in normal and cancerous tissues
Cellular incorporation of BrdU during S phase can be readily detected by anti-BrdU-specific antibodies following membrane permeabilization. Subsequent quantitation of cells positive for BrdU is possible using routine methods for cell analysis
  • Measures labeling indices of different cell populations
  • Improved specific staining with minimal background
  • Designed antigen retrieval solution unmask antigenic sites and preserve tissue morphology
  • Enables study of proliferation state of phenotypically defined cells within the micro-environment of tissues
  • Reaction: 50 slides
Flow Cytometry, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Research use only