Learn More
Brilliant Stain Buffer, BD
Catalog No. BDB563794 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
Catalog No. Quantity
BDB563794 100 Tests
1 options
Catalog No. BDB563794 Supplier BD Biosciences Supplier No. 563794
Add to Cart
Add to Cart
Designed to complement multicolor flow cytometry experiments. Brilliant Stain Buffer is a buffer for the immunofluorescent staining of cells.
  • Compatible with the use of other fluorescent staining reagents conjugated with traditional fluorochromes
  • Research Use Only
TRUSTED_SUSTAINABILITY
Specifications
Quantity 100 Tests