Learn More
BD Biocoat™ Disposable Syringe, Non-Sterile, Bulk Pack
Catalog No. 148268 Shop All Waters Corporation Products
Change view
Click to view available options
Volume (Metric):
20 mL
Catalog No. Volume (Metric)
14-826-8 20 mL
1 options
Catalog No. 14-826-8 Supplier BD Supplier No. 301032
Add to Cart
Add to Cart
For your general-purpose applications
  • 20mLCapacity
  • Latex-free, disposable polypropylene syringes 
  • Come nonsterilized in bulk packages and can be autoclaved.
  • Slip-tip
Order Info
Needles and shield tips not included
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Material Plastic
Product Type Syringe
Disposable Single-use
Volume (Metric) 20 mL
Connection Type Slip Tip
Graduated Yes
Sterility Non-sterile