Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Butterfields Buffer Solution

Catalog No. b292955 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
For Use With (Application):
Microbial blank dilutions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
B2-929-55 Microbial blank dilutions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. B2-929-55 Supplier BD Supplier No. 292955
This item is not returnable. View return policy

For isolation and/or differentiation of microorganisms from sampls or specimens

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 4°C
Additive Butterfield Buffer Solution
For Use With (Application) Microbial blank dilutions
Packaging Type Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.