Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD C5a, Recombinant mouse

Catalog No. BDB622597 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
10 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB622597 10 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB622597 Supplier BD Supplier No. 622597
Add to Cart
Add to Cart

Bioactive cleavage recombinant protein released from plasma component C5 during complement activation

  • Anaphylatoxin and potent pro-inflammatory agent
  • Responsible for mediation of variety of cellular immune responses
  • Purified by affinity chromatography
  • Storage buffer: Lyophilized in an aqueous buffered solution containing BSA and ProClin 150

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Type Purified Recombinant Mouse C5a
Quantity 10 Tests
Form Lyophilized
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.