Learn More
BD Pharmingen™ Calcein Blue AM Fluorescent Dye
Catalog No. BDB564060 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
BDB564060 1 mg
1 options
Catalog No. BDB564060 Supplier BD Supplier No. 564060
Add to Cart
Add to Cart
Calcein Blue AM is used for labeling live cells that can be detected and further analyzed by flow cytometry.
  • Fluoresent dye for labeling live cells.
TRUSTED_SUSTAINABILITY
Specifications
Description Calcein AM fluorescent dye
For Use With (Application) Flow Cytometry
Quantity 1 mg
WARNING: Reproductive Harm - www.P65Warnings.ca.gov