Learn More
BD Cell Analysis 3P 3 Beads, for use with FACSComp or AutoCOMP software, 25 tests
Supplier: BD Cell Analysis 3P 340486
BD CaliBRITE beads are designed for use with FACSComp or AutoCOMP software and the FACS family of flow cytometers (FACSCalibur, FACSort, FACScan, and FACStrak). The beads are used to adjust instrument settings, set fluorescence compensation, and check instrument sensitivity. Daily use is recommended for monitoring instrument performance over time. CaliBRITE beads are for in vitro diagnostic use. Store CaliBRITE beads at 2 deg. to 8 deg.C and protected from direct light. Do not use after the expiration date shown on the label. Store Bead Dilution Buffer at 2 deg. to 8 deg.C. After dilution, CaliBRITE bead suspensions prepared in Bead Dilution Buffer are stable for 8 hours at 2 deg. to 8 deg.C or for 1 hour at 20-25 deg.C. After dilution, CaliBRITE bead suspensions prepared in FACSFlow sheath fluid are stable for 8 hours at 2 deg. to 8 deg.C or, if PerCP beads are included, for 1 hour at 2 deg. to 8 deg.C.