Learn More
BD Cell Analysis 3P APC Beads, for use with FACSComp or AutoCOMP software, 25 tests
Catalog No. 50620176
Encompass
BD CaliBRITE beads are designed for use with FACSComp or AutoCOMP software and the FACS family of flow cytometers (FACSCalibur, FACSort, FACScan, and FACStrak). The beads are used to adjust instrument settings, set fluorescence compensation, and check instrument sensitivity. Daily use is recommended for monitoring instrument performance over time. CaliBRITE beads are for in vitro diagnostic use. 25 tests
Catalog No. 50-620-176 Supplier BD Cell Analysis 3P Supplier No. 340487
BD CaliBRITE beads are designed for use with FACSComp or AutoCOMP software and the FACS family of flow cytometers (FACSCalibur, FACSort, FACScan, and FACStrak). The beads are used to adjust instrument settings, set fluorescence compensation, and check instrument sensitivity. Daily use is recommended for monitoring instrument performance over time. CaliBRITE beads are for in vitro diagnostic use. 25 tests