Learn More
Description
Includes
Capture antibody coated microplate, detection antibody/enzyme conjugate (100x), standard (10x), wash buffer, TMB substrate, stop solution
For detection and quantitation of canine CRP in canine serum
- Reactivity: Dog
- Utilizes an antibody specific for canine CRP coated on a 96-well plate
Enzyme-linked immunosorbent assay
Specifications
Specifications
| Conjugate | HRP |
| Cross Reactivity | Dog |
| Includes | Capture Antibody coated microplate, detection Antibody/Enzyme Conjugate (100X), standard (10X), Wash Buffer, TMB Substrate, stop solution |
| Quantity | 1 Plate |
| Sample Type | Serum |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Detection and quantitation of Canine CRP in Canine Serum, ELISA |
| Product Type | Canine C-reactive Protein [CRP] ELISA kit |
Safety and Handling
Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.