Learn More
Andwin Scientific BD Cannula, Blunt, Plastic, Universal, Needleless Access
SDP
Catalog No. NC3994516
Encompass
BD Blunt Plastic Cannula is a universal cannula that can be used with all types of split septum IV injection sites and vials designed for needleless access. It provides simplified access to the Interlink IV Access System with smoother insertion and reduced penetration force.
  • Can be used with all types of split septum IV injection sites and vials
  • Designed for needleless access
  • Provides simplified access to Interlink IV Access System
  • Smoother insertion
  • Reduced penetration force
Catalog No. NC3994516 Supplier Andwin Scientific Supplier No. 12777124
BD Blunt Plastic Cannula is a universal cannula that can be used with all types of split septum IV injection sites and vials designed for needleless access. It provides simplified access to the Interlink IV Access System with smoother insertion and reduced penetration force.
  • Can be used with all types of split septum IV injection sites and vials
  • Designed for needleless access
  • Provides simplified access to Interlink IV Access System
  • Smoother insertion
  • Reduced penetration force