Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD CBA Mouse Immunoglobulin Isotyping Kit

Catalog No. BDB550026 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550026 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550026 Supplier BD Supplier No. 550026
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Heavy and light chain isotypes of mouse IgG1, IgG2a, IgG2b, IgG3, IgA, IgM, and IgE; reagents for capture of mouse IgG1, IgG2a, IgG2b, IgG3, IgA, IgM, and IgE; anti-kappa and anti-lambda detection reagents; standards; buffers; cytometer setup reagents

Combines a conventional immunoassay with the sensitivity of fluorescence via flow cytometry

  • Provides a capture surface for a specific protein and is analogous to an individually coated well in an ELISA plate
  • Broad dynamic range of fluorescence detection, combined with multiple analyte detection, allows for small sample volume, fewer sample dilutions, and substantially less time overall to establish the value of an unknown versus a conventional ELISA approach
  • Can be used to rapidly identify the heavy and light chain isotypes of a mouse monoclonal antibody in a single sample
  • Contains enough reagents to perform 100 tests

TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Cross Reactivity Mouse
Includes Heavy and light chain isotypes of Mouse IgG1, IgG2a, IgG2b, IgG3, IgA, IgM and IgE; reagents for capture of Mouse IgG1, IgG2a, IgG2b, IgG3, IgA, IgM and IgE, Anti-kappa and Anti-lambda detection reagents, standards, Buffers, cytometer setup reagents
Quantity 100 Tests
Content And Storage 2°C to 8°C
Form Liquid
For Use With (Application) Flow Cytometry
Product Type Mouse Immunoglobulin isotyping kit

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.