Learn More
Description
Includes
Reagents for capture and detection of IL-6, IL-10, MCP-1, IFN-γ, TNF, and IL-12p70; wash buffer; assay diluent; cytometer setup reagents
- Can be used to quantitatively measure Interleukin-6 (IL-6), Interleukin-10 (IL-10), Monocyte Chemoattractant Protein-1 (MCP-1), Interferon-γ (IFN-γ), Tumor Necrosis Factor (TNF), and Interleukin-12p70 (IL-12p70) protein levels in a single sample
- Detection reagent provided in the kit is a mixture of phycoerythrin (PE)-conjugated antibodies, which provides a fluorescent signal in proportion to the amount of bound analyte
- Provides sufficient reagents for 80 tests
For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications
Specifications
| Antigen | IFN-gamma |
| Conjugate | PE |
| Cross Reactivity | Mouse |
| Includes | Reagents for capture and detection of IL-6, IL-10, MCP-1, IFN-g, TNF and IL-12p70, Wash Buffer, assay diluent, cytometer setup reagents |
| Quantity | 80 Tests |
| Content And Storage | 2°C to 8°C |
| Form | Liquid |
| For Use With (Application) | Flow Cytometry |
| Product Type | Mouse inflammation kit |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.