Learn More
BD CBA Mouse Th1/Th2 Cytokine Kit
Catalog No. BDB551287 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
80 Tests
Catalog No. Quantity
BDB551287 80 Tests
1 options
Catalog No. BDB551287 Supplier BD Supplier No. 551287
Add to Cart
Add to Cart
Includes
Reagents for capture and detection of IL-2, IL-4, IL-5, IFN-γ, and TNF; standards; wash buffer; assay diluent; cytometer setup reagents
Optimized for analysis of physiologically relevant concentrations (pg/mL levels) of specific cytokine proteins in tissue culture supernatants and serum samples
BD CBA assays provide a method of capturing a soluble analyte or set of analytes with beads of known size and fluorescence, making it possible to detect analytes using flow cytometry.
  • Can be used to measure Interleukin-2 (IL-2), Interleukin-4 (IL-4), Interleukin-5 (IL-5), Interferon-γ (IFN-γ), and Tumor Necrosis Factor (TNF) protein levels in a single sample
  • Provides sufficient reagents for analysis of 80 samples
TRUSTED_SUSTAINABILITY
Specifications
Detects IL-2, IL-4, IL-5, IFN-g and TNF
For Use With (Application) For analysis of physiologically relevant concentrations (pg/mL levels) of specific Cytokine Proteins in Tissue Culture supernatants and Serum samples
Quantity 80 Tests
Content And Storage 2°C to 8°C
For research use only.