Learn More
BD CBA Mouse Th1/Th2/Th17 Cytokine Kit
Catalog No. BDB560485 Shop All Waters Corporation Products
Change view
Click to view available options
Quantity:
80 Tests
Catalog No. Quantity
BDB560485 80 Tests
1 options
Catalog No. BDB560485 Supplier BD Supplier No. 560485
Add to Cart
Add to Cart
Includes
Reagents for capture and detection of IL-2, IL-4, IL-6, IFN-γ, TNF, IL-17A, and IL-10; wash buffer; assay diluent; cytometer setup reagents
Optimized for analysis of physiologically relevant concentrations (pg/mL levels) of specific cytokine proteins in tissue culture supernatants and serum samples
BD CBA assays provide a method of capturing a soluble analyte or set of analytes with beads of known size and fluorescence, making it possible to detect analytes using flow cytometry.
  • Can be used to measure Interleukin-2 (IL-2), Interleukin-4 (IL-4), Interleukin-6 (IL-6), Interferon-γ (IFN-γ), Tumor Necrosis Factor (TNF), Interleukin-17A (IL-17A), and Interleukin-10 (IL-10) protein levels in a single sample
  • Provides sufficient reagents for 80 tests
TRUSTED_SUSTAINABILITY
Specifications
Detects IL-2, IL-4, IL-6, IFN-g, TNF, IL-17A and IL-10
For Use With (Application) For analysis of physiologically relevant concentrations (pg/mL levels) of specific Cytokine Proteins in Tissue Culture supernatants and Serum samples
Quantity 80 Tests
Content And Storage 2°C to 8°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For research use only.