Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD CD1d:Ig Recombinant Fusion Protein-Dimeric, Human

Catalog No. BDB557764 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557764 0.1 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557764 Supplier BD Supplier No. 557764
Add to Cart
Add to Cart

For studying Natural Killer T (NKT)-cell function by immunofluorescent staining

  • Format: Purified
  • Concentration: 0.5mg/ml
  • Isotype: Mouse IgG1, γ
  • Reactivity: Human
  • Storage buffer: Aqueous buffered solution containing ≤0.09% sodium azide

Flow Cytometry, Enzyme Linked Immunosorbent Assay

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5mg/mL
For Use With (Application) ELISA, Flow Cytometry
Formulation Aqueous Buffered solution containing ≤0.09% Sodium Azide
Name Human CD1d:Ig Recombinant Fusion Protein
Purification Method Affinity Chromatography, SDS-PAGE
Quantity 0.1 μg
Storage Requirements 4°C
Cross Reactivity Human
Recombinant Recombinant
Form Solution

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.