Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cell Cocktail

Catalog No. BDB558495 Shop All BD Biosciences Products
Change view
Click to view available options
Rat T, B, and NK lymphocyte Cell Cocktail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
BDB558495 Rat T, B, and NK lymphocyte Cell Cocktail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BDB558495 Supplier BD Supplier No. 558495
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Three-color reagent cocktail

  • Cocktail component: APC anti-Rat CD3, FITC anti-Rat CD45RA, PE anti-Rat CD161a
  • Components has three different clone: 1F4, OX-33, 10/78 respectively
  • Isotype: mIgM,mIgG1, mIgG1
  • Designed to identify rat T, B, and NK lymphocyte populations by direct immunofluorescent
  • Reactivity: Rat
  • Storage buffer: aqueous buffered solution containing BSA and ≤0.09% sodium azide

Flow cytometry

TRUSTED_SUSTAINABILITY

For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.