Learn More
BD Cell Cycle Kit
Catalog No. BDB558662 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
BDB558662 1 Kit
1 options
Catalog No. BDB558662 Supplier BD Supplier No. 558662
Includes
Alexa Fluor 488 Mouse anti-BrdU, Alexa Fluor™ 647 Rat anti-Histone H3, 5x Fixation Buffer, Perm Buffer III, Stain Buffer (FBS), BrdU, PBS (10X) concentrate, DNase, Hoechst 33342 solution
Testing of various time points and/or drug treatments to determine cell cycle kinetics
  • Utilizes antibodies that are directly conjugated with fluorochomes
  • Fluoresce in green (Alexa Fluor 488) and red (Alexa Fluor 647) channels
  • Allows for additional reagent in orange (Alexa Fluor 555) channel to be multiplexed with kit antibodies
Bioimaging
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Research use only