Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cell Enrichment Cocktail

Catalog No. BDB558569 Shop All BD Biosciences Products
Change view
Click to view available options
Product Type:
Naive CD4+ T Cell Cocktail
Naive CD8+ T Cell Cocktail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
BDB558569 Naive CD8+ T Cell Cocktail
BDB558521 Naive CD4+ T Cell Cocktail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB558569 Supplier BD Supplier No. 558569
Add to Cart
Add to Cart

Includes

Human Memory CD4+ T Cell Enrichment Cocktail, 5.0mL:

  • Biotin Mouse Anti-human CD8, clone SK1
  • Biotin Mouse Anti-human CD11b, clone ICRF44
  • Biotin Mouse Anti-human CD16, clone 3G8
  • Biotin Mouse Anti-human CD19, clone HIB19
  • Biotin Mouse Anti-human CD36, clone CB38
  • Biotin Mouse Anti-human CD41a, clone HIP8
  • Biotin Mouse Anti-human CD45RA, clone HI100
  • Biotin Mouse Anti-human CD56, clone B159
  • Biotin Mouse Anti-human CD123, clone 9F5
  • Biotin Mouse Anti-human CD235a (Glycophorin A), clone GA-R2 (HIR2)
  • Biotin Mouse Anti-human γδ TCR, clone B1
  • In aqueous buffered solution containing 0.09% Sodium Azide

BD IMag Streptavidin Particles Plus - DM, 7.5mL:

  • In aqueous buffered solution containing BSA™ and 0.09% Sodium Azide

For negative selection of untouched memory CD4+ helper T cells from peripheral blood

Contains biotinylated monoclonal antibodies that recognize antigens expressed on naive T cells, CD8+ T cells, B cells, NK cells, Γ/δ T cells, monocytes, DCs, granulocytes, platelets, and erythroid cells
Negative selection is performed to enrich for unlabeled memory cells. Labeled cells migrate towards magnet (positive fraction), leaving the unlabeled cells in suspension so they can be drawn off and retained (enriched fraction)

  • Contains biotinylated monoclonal antibodies that recognize antigens expressed on naive T cells, CD8+ T cells, B cells, NK cells, γ/δ T cells, monocytes, DCs, granulocytes, platelets, and erythroid cells
  • Avoids inadvertent activation of enriched memory helper T cells by using reagents that do not directly bind to those T cells
  • Simultaneously stains erythrocytes, platelets, and most leukocytes

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage In aqueous buffered solution containing BSA™ 0.09% sodium azide at 4°C
For Use With (Application) For negative selection of untouched memory CD4+ helper T cells from peripheral blood, magnetic cell separation
For Use With (Equipment) Flow cytometer
Includes Biotin mouse Anti-Human CD8, clone SK1, CD11b, clone ICRF44, CD16, clone 3G8, CD19, clone HIB19, CD36, clone CB38, CD41a, clone HIP8, CD45RA, clone HI100, CD56, clone B159, CD123, clone 9F5, CD235a (Glycophorin A), clone GA-R2 (HIR2), gd TCR, clone B1
Product Type Naive CD8+ T Cell Cocktail
Quantity 5 mL

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.