Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cell Lysis Buffer

Catalog No. BDB559759 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559759 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559759 Supplier BD Supplier No. 559759

missing translation for 'validMainframeMsg'

A convenient, ready-to-use non-ionic detergent based buffer that can be used for solubilizing cells needed for biochemical analysis

  • Composed of 10mM Tris-HCL (pH 7.5), 10mM NaH2PO4/NaHPO4, 130mM NaCl, 1% Triton X-100, 10mM PPi (sodiumnpyrophosphate)
  • Sterile, filtered
  • For use with the Caspase-3 Assay Kit and Caspase Enzyme Sets

TRUSTED_SUSTAINABILITY

Specifications

Formulation 10mM Tris pH 7.5, 130mM NaCl, 1% Trito n X-100™, 10mM NaF, 10mM NaPi, 10mM NaPPi
Quantity 50 mL
Format Liquid
Product Type Cell Lysis Buffer
For Use With (Application) Solubilizing cells needed for biochemical analysis of caspase-3 assay kit and caspase Enzyme sets
Content And Storage 4°C

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.