Learn More
Gibco™ Cell MAb Medium, animal component free
GREENER_CHOICE
Catalog No. CB220513 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1000 mL
Catalog No. Quantity
CB220513 1000 mL
1 options
Catalog No. CB220513 Supplier Gibco™ Supplier No. 220513
Add to Cart
Add to Cart
Produces yields higher than classical basal media with similar supplementation
Promotes an acceleration in the monoclonal antibody secretion rates of cells in a batch mode for performance in hollow fiber and stir tank systems.
  • Suitable for use in perfusion systems
  • Shown to increase purified monoclonal antibody yields by 5 to 25 times over classical basal media with similar supplementation
  • Reduce media consumption and labor cost, reduce requirements for media storage and preparation time, decrease labor required to perform continuous feeding and decrease contamination because of decreased handling of the culture
  • Concentration step is eliminated in most cases, and final bulk harvest is reduced, producing less material to run through the purification process
  • Only minimal adaptation is needed
  • Hybridomas grown in MAb Basal Medium can be supplemented in the same way as those grown in traditional basal media
  • MAb Serum-Free and Animal-Free are complete media and do not require serum supplementation; Basal Medium does require serum supplementation
TRUSTED_SUSTAINABILITY
Specifications
Product Type MAb Medium
With Additives Glutamine
Without Additives No Phenol Red, No Pluronic Acid, No Other Surfactants
Quantity 1000 mL
Content And Storage Storage conditions: 2°C to 8°C. Protect from light.