Learn More
Description
Includes
- Assay diluent
- Capture bead diluent
- Detection reagent diluent
- Denaturation buffer
- Wash buffer
- Instrument setup reagents
The buffers and instrument setup reagents provided in this kit have been optimized to measure total or phosphorylated proteins from denatured cell lysate sample.
- Contains all of the supporting reagents necessary to perform an assay using a BD CBA Cell Signaling Flex Set
- Provides sufficient reagents for the quantitative analysis of 100 samples and 10 instrument setup procedures
- Working assay range for most analytes in this system is 3.9 to 1,000 units/mL
For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| No. of Tests | 500 |
| Includes | 1 bottle 150mL assay diluent, 1 bottle 30mL capture bead diluent, 1 bottle 30mL detection reagent diluent, 1 bottle 150mL denatuRation buffer 5X, 1 bottle 650mL wash buffer, 1 vial 0.25mL instrument setup bead A1, 1 vial 0.25mL instrument setup bead A9 |
| Validated Application | RUO |
| Product Type | Cytometric Bead Array (CBA) Assay |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.