Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cell Signaling Master Buffer Kit

Catalog No. p-4003663 Shop All BD Biosciences Products
Change view
Click to view available options
No. of Tests:
100
500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Tests
BDB560005 100
BDB560006 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB560005 Supplier BD Supplier No. 560005

Includes

  • Assay diluent
  • Capture bead diluent
  • Detection reagent diluent
  • Denaturation buffer
  • Wash buffer
  • Instrument setup reagents

Employ particles with discrete fluorescence intensities to detect soluble analytes at very low concentrations

The buffers and instrument setup reagents provided in this kit have been optimized to measure total or phosphorylated proteins from denatured cell lysate sample.

  • Contains all of the supporting reagents necessary to perform an assay using a BD CBA Cell Signaling Flex Set
  • Provides sufficient reagents for the quantitative analysis of 100 samples and 10 instrument setup procedures
  • Working assay range for most analytes in this system is 3.9 to 1,000 units/mL

For research use only. Not for use in diagnostic or therapeutic procedures.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Cytometric Bead Array (CBA) Assay
Includes 1 bottle 30mL assay diluent, 1 bottle 5mL capture bead diluent, 1 bottle 5mL detection reagent diluent, 1 bottle 30mL denaturation buffer 5X, 1 bottle 130mL wash buffer, 1 vial 0.25mL instrument setup bead A1, 1 vial 0.25mL instrument setup bead A9
Content And Storage 2°C to 8°C
No. of Tests 100
Validated Application RUO
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.