Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Chek™ Group A Strep Test Kit

Catalog No. 14910125 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-910-125 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-910-125 Supplier BD Supplier No. 256031

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapid chromatographic immunoassay for the qualitative detection of Strep A antigen from throat swab specimens. Results in just five minutes.

  • Sensitivity: 95.1%
  • Specificity: 99.0%
  • Simple steps with easy-to-read results
  • Results in just five minutes
  • Reusable five-test workstation included in each kit
  • Built-in procedural controls in each individually packaged test strip
  • For in vitro diagnostic use
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 30°C
Clia Complexity Waived
Detectable Analytes Streptococci
Sample Type Throat Swab
Sensitivity 95.1%
Type Group A Streptococcus Test Kit
Quantity 25 Tests
Specificity 0.99
Test Time 5 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.