Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD CompBead Plus Anti-Rat Ig, κ/Negative Control Compensation Plus (7.5μm) Particles Set

Catalog No. BDB560499 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560499 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560499 Supplier BD Supplier No. 560499
Add to Cart
Add to Cart

Includes

  • Negative Control (PBS with 1% BSA)
  • Anti-Rat Ig, κ

Provides two types of particles, CompBead Plus Anti-Rat Ig, κ particles, which binds any rat κ light chain-bearing immunoglobulin, and CompBead Plus Negative Control (BSA) particles, which has no binding capacity

  • Mixed together with fluorochrome-conjugated rat antibody, CompBead Plus provides distinct positive and negative (background fluorescence) stained populations which can be used to set compensation levels manually or using instrument set-up software

TRUSTED_SUSTAINABILITY

Specifications

Applications Flow Cytometry
Quantity 1 Each
Regulatory Status RUO
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.