Learn More
BD Cytofix/Cytoperm™ Fixation/Permeabilization Solution Kit with BD GolgiStop™
Shop All Waters Corporation ProductsDescription
Includes
- Fixation/Permeabilization solution (125mL)
- BD Perm/Wash™ Buffer, 10x concentrate containing Fetal Bovine Serum (FBS) and saponin (100mL)
- Protein transport inhibitor BD GolgiStop™ (containing Monensin)
The ex vivo addition of GolgiStop to in vitro- or in vivo -stimulated cells blocks their intracellular transport processes. This results in the accumulation of most cytokine proteins in the Golgi complex and thereby enhances cytokine staining signals.
- GolgiStop reagent is provided for treating up to 1L of cell culture at a cell density of up to 2 × 106 cells/mL
Specifications
Specifications
| Quantity | 250 Tests |
| Product Type | Fixation/Permeabilization Reagent |
| Includes | 125 mL fixation/permeabilization solution, perm/Wash™ buffer, fetal bovine serum (FBS) 10X and 100 mL saponin, protein transport inhibitor BD GolgiStop™ |
| No. of Tests | 250 |
| Regulatory Status | RUO |
| Content And Storage | 4°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.