Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytofix/Cytoperm Fixation/Permeabilization Solution Kit with GolgiPlug™

Catalog No. bdb555028 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
250 Tests
Catalog No. Quantity
BDB555028 250 Tests
1 options

Catalog No. BDB555028

Supplier: BD 555028

Only null left
Add to Cart
Add to Cart

Includes

Fixation/Permeabilization solution (125mL), BD Perm/Wash™ Buffer, 10X concentrate containing Fetal Bovine Serum (FBS) and saponin (100mL), and protein transport inhibitor BD GolgiPlug (containing Brefeldin A)

For immunofluorescent staining of cytokines

  • Offers three kits to simplify fixation and permeabilization of cells
  • For immunofluorescent staining of intracytoplasmic cytokines

All three kits described in this product manual contain Fixation/Permeabilization solution containing 4% paraformaldehyde, and either BD GolgiStop™ protein transport inhibitor containing 99.9% ethanol, or BD GolgiPlug protein transport inhibitor containing 99.9% dimethylsulfoxide and 0.1% brefeldin A. The following warnings and precautions apply to the aforementioned chemicals.
R 36/38: Irritating to eyes and skin
S3: Keep in a cool place
S26: In case of contact with eyes, rinse immediately with water and seek medical advise
S28: After contact with skin, wash immediately with plenty of water
R40: Limited evidence of a carcinogenic effect
S36/37: Wear suitable protective clothing and gloves

Flow Cytometry, Intracellular staining

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) For immunofluorescent staining of cytokines
Quantity 250 Tests
Content And Storage 2°C to 8°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.