Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Functional Bead Conjugation Buffer Set

Catalog No. bdb558556 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
15 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558556 15 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.
Catalog No. BDB558556 Supplier BD Supplier No. 558556
Only null left
Add to Cart
Add to Cart
Sodium azide yields highly toxic hydrazoic acid under acidic conditions. Dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing.

Includes

Coupling buffer and storage buffer

Used to covalently link watersoluble proteins to surface of color-coded functional beads

  • Contains supporting reagents for BD™ CBA Functional Bead system
  • Simple conjugation procedure completed within 4 hours
  • Conjugation reaction requires less then 100μg of protein at a concentration of 1mg/mL
  • Size: 15 tests

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RUO
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Functional Bead Conjugation Buffer Set
For Use With (Equipment) BD™ CBA functional bead system
Includes Coupling buffer and storage buffer and supporting reagents
No. of Tests 15
Quantity 15 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.