Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Functional Beads

Catalog No. BDB558578 Shop All BD Biosciences Products
Change view
Click to view available options
Product Type:
Functional Bead (A4)
Functional Bead (A5)
Functional Bead (A6)
Functional Bead (A7)
Functional Bead (A8)
Functional Bead (A9)
Functional Bead (B4)
Functional Bead (B5)
Functional Bead (B6)
Functional Bead (B7)
Functional Bead (B8)
Functional Bead (C4)
Functional Bead (C6)
Functional Bead (C8)
Functional Bead (C9)
Functional Bead (D4)
Functional Bead (D5)
Functional Bead (D6)
Functional Bead (D7)
Functional Bead (D8)
Functional Bead (E4)
Functional Bead (E5)
Functional Bead (E6)
Functional Bead (E7)
Functional Bead (E8)
Functional Bead (E9)
26 product options available for selection
Product selection table with 26 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
BDB558578 Functional Bead (A4)
BDB558580 Functional Bead (B7)
BDB558582 Functional Bead (C6)
BDB558581 Functional Bead (B8)
BDB558583 Functional Bead (D5)
BDB558579 Functional Bead (A9)
BDB558584 Functional Bead (E4)
BDB558586 Functional Bead (B5)
BDB558647 Functional Bead (C4)
BDB558650 Functional Bead (C9)
BDB558656 Functional Bead (D8)
BDB560031 Functional Bead (A5)
BDB560032 Functional Bead (A6)
BDB560035 Functional Bead (B4)
BDB560034 Functional Bead (A8)
BDB560036 Functional Bead (D4)
BDB560037 Functional Bead (E5)
BDB560040 Functional Bead (E8)
BDB558585 Functional Bead (E9)
BDB558645 Functional Bead (B6)
BDB558649 Functional Bead (C8)
BDB558654 Functional Bead (D6)
BDB558655 Functional Bead (D7)
BDB560033 Functional Bead (A7)
BDB560038 Functional Bead (E6)
BDB560039 Functional Bead (E7)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 26 options available.
26 options
Catalog No. BDB558578 Supplier BD Supplier No. 558578
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Color-coded 7.5μm polystyrene beads that can be classified using a flow cytometer

The Functional Beads can be used in conjunction with the BD CBA Functional Bead Conjugation Buffer Set (Mfr. No. 558556) to covalently link water-soluble proteins to the surface of the beads via a sulfo-SMCC chemistry. The beads, once linked to a protein, can be used as a capture matrix for analytes of interest.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RUO
Content And Storage 4°C
Contents Aqueous buffered solution containing ≤0.09% sodium azide
Description Functional Bead (A4)
For Use With (Application) Flow Cytometry
Material Polystyrene
Product Type Functional Bead (A4)
For Use With (Equipment) Flow cytometer
No. of Tests 6000
Volume (Metric) 0.9 mL
Diameter (Metric) 7.5 μm
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.