Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Human Angiogenin; Flex Set, Bead C4

Catalog No. BDB558328 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558328 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558328 Supplier BD Supplier No. 558328
Add to Cart
Add to Cart

Includes

Standard, PE Detection Reagent, Capture Bead C4

For measuring human angiogenin in serum, plasma and cell culture supernatant samples

  • Capture bead and PE detection reagent formulated to 50x concentration
  • Includes lyophilized standard

TRUSTED_SUSTAINABILITY

Specifications

Antigen Angiogenin
Cross Reactivity Human
Includes 1 Vial capture bead 50X, 1 vial detection reagent 50X, 2 vials standard (lyophilized)
Quantity 100 Tests
Sample Type Serum, Plasma, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Flow cytometry, for measuring Human angiogenin
Product Type Human Angiogenin flex set (bead C4)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.