Learn More
BD Cytometric Bead Array (CBA) Human CD121a (IL-1 RI); Flex Set, Bead B6
Shop All BD Biosciences ProductsDescription
Includes
Human CD121a (IL-1 RI), Standard; PE Detection Reagent; Capture Bead B6.
- Bead-based immunoassay
- Also known as IL-1 Receptor type-1
- Human reactivity determined by testing samples with BD CBA Human Soluble CD121a (IL-1 RI) Flex Set
Specifications
Specifications
| Antigen | CD121alpha (IL-1 RI) |
| Cross Reactivity | Human |
| Includes | Human CD121a (IL-1 RI), standard, PE detection reagent, capture bead B6 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| Form | Buffer Solution |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human soluble CD121a (IL-1 RI) flex set (bead B6) |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.