Learn More
BD Cytometric Bead Array (CBA) Human CD121b (IL-1 RII); Flex Set, Bead B7
Shop All BD Biosciences ProductsDescription
Includes
Human CD121b (IL-1 RII), Standard; PE Detection Reagent; Capture Bead B7.
- Also known as IL-1 Receptor Type-II
- Bead-based immunoassay
- Human reactivity determined by testing samples with BD CBA Human Soluble CD121b Flex Set
Specifications
Specifications
| Antigen | CD121beta (IL-1 RII) |
| Cross Reactivity | Human |
| Includes | Human CD121b (IL-1 RII), standard, PE detection reagent, capture bead B7 |
| Quantity | 100 Tests |
| Content And Storage | 4°C |
| Form | Lyophilized |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human CD121b (IL-1 RII) flex set (bead B7) |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.