Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Human CD178 (FASL); Flex Set, Bead C6

Catalog No. BDB558330 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558330 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558330 Supplier BD Supplier No. 558330
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Human CD178 (FASL); Standard; Human CD178 (FASL); PE Detection Reagent; Human CD178 (FASL); Capture Bead C6

For detecting soluble human Fas Ligand in serum, plasma, and cell culture samples

The BD CBA Human CD178 (FASL) Flex Set is a bead-based immunoassay capable of measuring soluble human Fas Ligand, also known as soluble CD178, in serum, plasma, and cell culture supernatant samples. Human reactivity was determined by testing samples with the BD CBA Human CD178 (Fas Ligand) Flex Set.

  • Reactivity: Human
  • Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
  • Standard component is lyophilized
  • When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
  • Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD178 (FASL)
Cross Reactivity Human
Includes Human CD178 (FASL), standard, Human CD178 (FASL), PE detection reagent, Human CD178 (FASL), capture bead C6
Quantity 100 Tests
Sample Type Serum, Plasma, Cell Culture Supernatant
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Human CD178 (Fas Ligand) flex set (bead C6)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.