Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Cytometric Bead Array (CBA) Human CD62L (L-Selectin); Flex Set, Bead D8

Catalog No. BDB560420 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560420 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Highly toxic, dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing
Catalog No. BDB560420 Supplier BD Supplier No. 560420
Add to Cart
Add to Cart
Highly toxic, dilute azide compounds in running water before discarding to avoid accumulation of potentially explosive deposits in plumbing

Includes

Human CD62L (L-Selectin); Standard; Human CD62L (L-Selectin); PE Detection Reagent; Human CD62L (L-Selectin); Capture Bead D8

Determines human reactivity by testing samples with BD CBA Human Soluble CD62L (L-Selectin) Flex Set

The BD CBA Human CD62L (L-Selectin) Flex Set is a bead-based immunoassay capable of measuring human soluble CD62L (L-Selectin) in serum, plasma and cell culture supernatant samples

  • Reactivity: QC Tested: Human
  • Bead Position: D8
  • Limit of detection: 28.6pg/mL

With BD CBA human soluble protein master buffer kit (Cat. No. 558264, 100 tests, or 558265, 500 tests), flow cytometer, and FCAP Array™ software

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD62L
Cross Reactivity Human
Includes Human CD62L (L-Selectin), standard, Human CD62L (L-Selectin), PE detection reagent, Human CD62L (L-Selectin), capture bead D8
Quantity 100 Tests
Content And Storage 4°C
For Use With (Application) Flow Cytometry
Product Type Human CD62L (L-Selectin) flex set (bead D8)

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.