Learn More
BD Cytometric Bead Array (CBA) Human CD62P (P-Selectin); Flex Set, Bead D7
Shop All BD Biosciences ProductsDescription
Includes
Human CD62P (P-Selectin); Standard; Human CD62P (P-Selectin); PE Detection Reagent; Human CD62P (P-Selectin); Capture Bead D7
Human reactivity was determined by testing samples with the BD CBA Human Soluble CD62P (P-Selectin) Flex Set.
- Format: Purified
Specifications
Specifications
| Antigen | CD62P (P-Selectin) |
| Conjugate | PE |
| Cross Reactivity | Human |
| Includes | Capture bead, detection reagent and standard |
| Quantity | 100 Tests |
| Form | Solid |
| For Use With (Application) | Flow Cytometry |
| Product Type | Human CD62P (P-Selectin) flex set (bead D7) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.